Integrity Testers
Various regulatory guidelines require integrity testing of sterilizing filters used in the processing of sterile solutions, such as large volume parenterals (LVPs) and small volume parenterals (SVPs).* With many biopharmaceutical manufacturers now testing filters before and after use, there is a need for non-destructive test methods that are fast, reliable, and repeatable. The Integritest® 5 automated test instrument meets this need by enabling non-destructive integrity testing to be performed inline or offline on filtration devices, with the flexibility to add a paired Exact-Air™ II water-based test unit for hydrophobic filters.
*EMA Annex 1; FDA Aseptic Processing cGMP guidance (2004)
Integritest® 5 automated test instrument
Our latest model since introducing the first commercial automated integrity test instruments more than 25 years ago, the Integritest® 5 uses pressure decay measurements to determine filter integrity. Along with a system volume measurement, one pressure decay measurement is taken to determine the relevant flowrate for diffusion and HydroCorr™ tests. The Tangent Method is used to determine a filter bubble point from a series of pressure decays performed at increasing transmembrane pressures. Optimum pressures at which to take measurements are based on test parameters and ongoing readings, reducing the number of measurements needed to reliably map the filter’s profile and return an accurate result.
Key features include:
- Improved test algorithm for faster, simpler testing
- Intuitive software with help screens for every step
- Flexible displays, run information, and test collections to suit your process
- Networking functions such as multi-unit management and synchronization
Exact-Air™ II automatic water-based test unit for hydrophobic filters
Based on Remote Volume Technology, the Exact-Air™ II System is an automatic integrity test unit with an intermediate volume located between the filter to be tested and the testing device. Paired with the Integritest® 5 tester, it improves test accuracy and reproducibility under extremely critical temperature conditions.
By using the HydroCorr™ test method, a highly sensitive water-based integrity test for hydrophobic membrane filters, the Exact-Air™ II System offers users a unique opportunity to improve reliability, accuracy, reproducibility, and frequency of air filter testing, even under challenging operating conditions.
Key features include:
- User-friendly software with touch screen display
- Control of filling, testing, draining, and cleaning-in-place operations
- Ease of validation
- Bubble point and diffusion testing on hydrophilic filters
Related Product Resources
Tech Brief: Bubble Point Testing with Integritest® 5
Tech Brief: Millipak® and Millidisk® Barrier Filters
Data Sheet: Exact-Air™ II Automatic, Water-based
Integrity Testing Option
Data Sheet: Millidisk® Barrier and Millipak® Barrier Filters
White Paper: Establishing Product Specific Bubble Point Specifications for Sterilizing-grade Filters
Application Note: Integrity Test Troubleshooting – Beyond Rewet and Retest