Merck
Milli-Q® EQ 7008 Ultrapure and Pure Water Purification System The Milli-Q®water system produces ultrapure (Type 1) and pure (RO, Type 3) water from tap water with a production flow rate up to 8 L/hour
Milli-Q® EQ 7008 Ultrapure and Pure Water Purification System
Milli-Q® EQ 7008 Ultrapure and Pure Water Purification System The Milli-Q®water system produces ultrapure (Type 1) and pure (RO, Type 3) water from tap water with a production flow rate up to 8 L/hour
Synonim(y):
Milli-Q® EQ 7008/16 Ultrapure and Pure Water Purification System
Products
Kategoria produktu
Marka
Linia produktu
Poziom jakości
Dzienne zużycie wody (l/dzień)
Wielkość opakowania
Jakość wody zasilającej
Jakość wody