Merck
SIAL0639
Sigma-Aldrich
Sigma® 25 cm cell culture flask
cap, vented, tissue-culture treated, sterile, pack of 10 × 20 ea
About This Item
Kod UNSPSC:
41104900
NACRES:
NB.22
Pozwól nam pomóc
Materiały
canted neck
polystyrene
sterylność
sterile
Właściwości
cap vented
graduations
opakowanie
pack of 10 × 20 ea
maksymalna pojemność
60 mL
powierzchnia
25 cm2
pojemność robocza
5-7.5 mL
typ wiązania
Tissue Culture (TC)-treated surface
Opis ogólny
Available in a variety of sizes, designs and cap styles to meet your needs.
  • Manufactured from optically clear virgin polystyrene
  • Treated for optimal cell attachment
  • Printed with lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation and certified nonpyrogenic
Informacje prawne
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Still not finding the right product?
Explore all of our products under Sigma® 25 cm cell culture flask
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)