
Yttria stabilized zirconia

Synonym(s):
YSZ, Yttria stabilized zirconia, Yttrium stabilized zirconium oxide
Linear Formula:
ZrO2 · Y2O3
CAS Number:
Molecular Weight:
349.03
Products
Product Category
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing