Merck
HPLC Analysis of 1-(1-Naphthyl)ethylamine Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of 1-(1-Naphthyl)ethylamine Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] methanol; [B] 20 mM ammonium acetate; (90:10, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
1 mgl/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC