Merck
  • FXR1 regulates transcription and is required for growth of human cancer cells with
FXR1 regulates transcription and is required for growth of human cancer cells with
eLife (2017-08-03)
Yichao Fan, Jiao Yue, Mengtao Xiao, Han Han-Zhang, Yao Vickie Wang, Chun Ma, Zhilin Deng, Yingxiang Li, Yanyan Yu, Xinghao Wang, Shen Niu, Youjia Hua, Zhiping Weng, Peter Atadja, En Li, Bin Xiang
ABSTRACT
Tumor suppressor p53 prevents cell transformation by inducing apoptosis and other responses. Homozygous
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-FXR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)