Merck
  • Stable ICG-loaded upconversion nanoparticles: silica core/shell theranostic nanoplatform for dual-modal upconversion and photoacoustic imaging together with photothermal therapy.
Stable ICG-loaded upconversion nanoparticles: silica core/shell theranostic nanoplatform for dual-modal upconversion and photoacoustic imaging together with photothermal therapy.
Scientific reports (2017-11-18)
Ruichan Lv, Depeng Wang, Liyang Xiao, Guanying Chen, Jun Xia, Paras N Prasad
ABSTRACT
We report here the design and multiple functions of a new hierarchical nanotheronostic platform consisting of an upconversion nanoparticle (UCNP) core: shell with an additional mesoporous silica (mSiO
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Sodium trifluoroacetate, 98%