Merck
  • Interplay between protein glycosylation pathways in Alzheimer's disease.
Interplay between protein glycosylation pathways in Alzheimer's disease.
Science advances (2017-09-21)
Moran Frenkel-Pinter, Merav Daniel Shmueli, Chen Raz, Michaela Yanku, Shai Zilberzwige, Ehud Gazit, Daniel Segal
ABSTRACT
Deviations from the normal nucleoplasmic protein O-GlcNAcylation, as well as from normal protein sialylation and N-glycosylation in the secretory pathway, have been reported in Alzheimer's disease (AD). However, the interplay between the cytoplasmic protein O-GlcNAcylation and the secretory N-/O-glycosylation in AD has not been described. We present a comprehensive analysis of the N-, O-, and O-GlcNAc-glycomes in AD-affected brain regions as well as in AD patient serum. We detected marked differences in levels of glycan involved in both protein O-GlcNAcylation and N-/O-glycosylation between patients and healthy individuals and revealed brain region-specific glycosylation-related pathology in patients. These alterations are not general for other neurodegenerative conditions, such as frontotemporal dementia and corticobasal degeneration. The alterations in the AD glycome in the serum could potentially lead to novel glyco-based biomarkers for AD progression. Strikingly, negative interrelationship was found between the pathways of protein O-GlcNAcylation and N-/O-glycosylation, suggesting a novel intracellular cross-talk.
Sigma-Aldrich
Trimethylboron, electronic grade, ≥98.35%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate