Merck
  • Mathematical modelling and evaluation of performance of cuboid packed-bed devices for chromatographic separations.
Mathematical modelling and evaluation of performance of cuboid packed-bed devices for chromatographic separations.
Journal of chromatography. A (2017-08-13)
Raja Ghosh, Guoqiang Chen
ABSTRACT
In a recent paper, box-shaped or cuboid packed bed devices have been proposed as alternative to columns for chromatographic separations. We first propose a mathematical model for residence time distribution in such devices. Based on it, we examine factors likely to affect separation performance, and verify the predictions of our mathematical model by conducting tracer experiments. We then compare the performance of two commercial columns with their respective equivalent cuboid packed-bed devices, i.e. containing the same chromatographic media, and having the same bed-height and bed-volume. Parameters compared include the number of theoretical plates, attributes of flow-through and eluted protein peaks, and resolution in model binary protein separations. For each of these metrics examined, the cuboid packed-bed device outperformed its equivalent commercial column. Other potential advantages likely to be gained from using a cuboid packed-bed instead of a column are elucidated.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein