Merck
  • Miconazole protects blood vessels from MMP9-dependent rupture and hemorrhage.
Miconazole protects blood vessels from MMP9-dependent rupture and hemorrhage.
Disease models & mechanisms (2017-02-06)
Ran Yang, Yunpei Zhang, Dandan Huang, Xiao Luo, Liangren Zhang, Xiaojun Zhu, Xiaolin Zhang, Zhenming Liu, Jing-Yan Han, Jing-Wei Xiong
ABSTRACT
Hemorrhagic stroke accounts for 10-15% of all strokes and is strongly associated with mortality and morbidity worldwide, but its prevention and therapeutic interventions remain a major challenge. Here, we report the identification of miconazole as a hemorrhagic suppressor by a small-molecule screen in zebrafish. We found that a hypomorphic mutant
Sigma-Aldrich
Decamethylcyclopentasiloxane, 97%
Sigma-Aldrich
Anti-MMP9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Triciribine hydrate, ≥97% (HPLC)