Merck
  • Identification of natural products with neuronal and metabolic benefits through autophagy induction.
Identification of natural products with neuronal and metabolic benefits through autophagy induction.
Autophagy (2016-10-30)
Yuying Fan, Nan Wang, Altea Rocchi, Weiran Zhang, Robert Vassar, Yifa Zhou, Congcong He
ABSTRACT
Autophagy is a housekeeping lysosomal degradation pathway important for cellular survival, homeostasis and function. Various disease models have shown that upregulation of autophagy may be beneficial to combat disease pathogenesis. However, despite several recently reported small-molecule screens for synthetic autophagy inducers, natural chemicals of diverse structures and functions have not been included in the synthetic libraries, and characterization of their roles in autophagy has been lacking. To discover novel autophagy-regulating compounds and study their therapeutic mechanisms, we used analytic chemistry approaches to isolate natural phytochemicals from a reservoir of medicinal plants used in traditional remedies. From this pilot plant metabolite library, we identified several novel autophagy-inducing phytochemicals, including Rg2. Rg2 is a steroid glycoside chemical that activates autophagy in an AMPK-ULK1-dependent and MTOR-independent manner. Induction of autophagy by Rg2 enhances the clearance of protein aggregates in a cell-based model, improves cognitive behaviors in a mouse model of Alzheimer disease, and prevents high-fat diet-induced insulin resistance. Thus, we discovered a series of autophagy-inducing phytochemicals from medicinal plants, and found that one of the compounds Rg2 mediates metabolic and neurotrophic effects dependent on activation of the autophagy pathway. These findings may help explain how medicinal plants exert the therapeutic functions against metabolic diseases.
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Insulin solution from bovine pancreas, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Earle′s Balanced Salt Solution 10x, Without sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein