Merck
  • Energy metabolic state in hypothermically stored boar spermatozoa using a revised protocol for efficient ATP extraction.
Energy metabolic state in hypothermically stored boar spermatozoa using a revised protocol for efficient ATP extraction.
Biology open (2016-09-11)
Quynh Thu Nguyen, Ulrike Wallner, Marion Schmicke, Dagmar Waberski, Heiko Henning
ABSTRACT
Mammalian spermatozoa utilize ATP as the energy source for key functions on the route to fertilization. ATP and its precursor nucleotides ADP and AMP are regularly investigated in sperm physiology studies, mostly by bioluminescence assays. Assay results vary widely, mainly due to different efficiencies in nucleotide extraction and prevention of their enzymatic degradation. Here, we describe a revised, validated protocol for efficient phosphatase inhibition and adenine nucleotide extraction resulting in consistently high ATP concentrations exceeding previously reported values for boar spermatozoa up to 20-fold. The revised assay is applicable for determining ATP concentrations and adenylate energy charge in extracts from fresh and frozen samples, thereby allowing simultaneous assessment of semen samples from long-term storage experiments. After validation, the assay was applied to liquid-preserved boar spermatozoa stored at 17°C and 5°C for 24 and 72 h. Cooling to 5°C, but not storage duration, reduced ATP concentration in spermatozoa (P<0.05), which was accompanied by the appearance of AMP and ADP in the preservation medium. ATP and energy charge were highly correlated to the proportion of membrane-intact spermatozoa, supporting the idea of nucleotides leaking through disrupted membranes in cold-shocked cells. The present assay allows highly standardized studies of energy metabolism in spermatozoa.
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) assay mix dilution buffer, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) assay mix, lyophilized powder