Merck
A New Member of the Graphene Family: Graphene Acid.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-10-22)
Ondřej Jankovský, Michal Nováček, Jan Luxa, David Sedmidubský, Vlastimil Fila, Martin Pumera, Zdeněk Sofer
ABSTRACT
A new member of the family of graphene derivatives, namely, graphene acid with a composition close to C
Sigma-Aldrich
Barium nitrate, AR, ≥99.5%