Merck
  • Phosphorus and Halogen Co-Doped Graphene Materials and their Electrochemistry.
Phosphorus and Halogen Co-Doped Graphene Materials and their Electrochemistry.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-09-09)
Lu Wang, Zdenek Sofer, Radek Zboril, Klara Cepe, Martin Pumera
ABSTRACT
Doping of graphene materials with heteroatoms is important as it can change their electronic and electrochemical properties. Here, graphene is co-doped with n-type dopants such as phosphorus and halogen (Cl, Br, I). Phosphorus and halogen are introduced through the treatment of graphene oxide with PX
Sigma-Aldrich
Barium nitrate, AR, ≥99.5%