Merck
  • Multiplexed In-cell Immunoassay for Same-sample Protein Expression Profiling.
Multiplexed In-cell Immunoassay for Same-sample Protein Expression Profiling.
Scientific reports (2015-09-04)
Jing Shang, Pavel Zrazhevskiy, Nadia Postupna, C Dirk Keene, Thomas J Montine, Xiaohu Gao
ABSTRACT
In-cell immunoassays have become a valuable tool for protein expression analysis complementary to established assay formats. However, comprehensive molecular characterization of individual specimens has proven challenging and impractical due to, in part, a singleplex nature of reporter enzymes and technical complexity of alternative assay formats. Herein, we describe a simple and robust methodology for multiplexed protein expression profiling on the same intact specimen, employing a well-characterized enzyme alkaline phosphatase for accurate quantification of all targets of interest, while overcoming fundamental limitations of enzyme-based techniques by implementing the DNA-programmed release mechanism for segregation of sub-sets of target-bound reporters. In essence, this methodology converts same-sample multi-target labeling into a set of isolated singleplex measurements performed in a parallel self-consistent fashion. For a proof-of-principle, multiplexed detection of three model proteins was demonstrated on cultured HeLa cells, and two clinically-relevant markers of dementia, β-amyloid and PHF-tau, were profiled in formalin-fixed paraffin embedded brain tissue sections, uncovering correlated increase in abundance of both markers in the "Alzheimer's disease" cohort. Featuring an analytically powerful yet technically simple and robust methodology, multiplexed in-cell immunoassay is expected to enable insightful same-sample protein profiling studies and become broadly adopted in biomedical research and clinical diagnostics.
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, powder
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, Vetec, reagent grade, ≥99%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Formaldehyde solution, LR, contains 5-8% methanol as stabilizer, 37-41 % (w/v)
Sigma-Aldrich
Formaldehyde solution, AR, contains 5-8% methanol as stabilizer, 37-41 % (w/v)
Sigma-Aldrich
Dodecyltrimethylammonium chloride, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Dodecyltrimethylammonium chloride, ≥99.0% (AT)