Merck
  • Anti-CSF-1 treatment is effective to prevent carcinoma invasion induced by monocyte-derived cells but scarcely by microglia.
Anti-CSF-1 treatment is effective to prevent carcinoma invasion induced by monocyte-derived cells but scarcely by microglia.
Oncotarget (2015-06-23)
Eva Rietkötter, Annalen Bleckmann, Michaela Bayerlová, Kerstin Menck, Han-Ning Chuang, Britta Wenske, Hila Schwartz, Neta Erez, Claudia Binder, Uwe-Karsten Hanisch, Tobias Pukrop
ABSTRACT
The mononuclear phagocytic system is categorized in three major groups: monocyte-derived cells (MCs), dendritic cells and resident macrophages. During breast cancer progression the colony stimulating factor 1 (CSF-1) can reprogram MCs into tumor-promoting macrophages in the primary tumor. However, the effect of CSF-1 during colonization of the brain parenchyma is largely unknown. Thus, we analyzed the outcome of anti-CSF-1 treatment on the resident macrophage population of the brain, the microglia, in comparison to MCs, alone and in different in vitro co-culture models. Our results underline the addiction of MCs to CSF-1 while surprisingly, microglia were not affected. Furthermore, in contrast to the brain, the bone marrow did not express the alternative ligand, IL-34. Yet treatment with IL-34 and co-culture with carcinoma cells partially rescued the anti-CSF-1 effects on MCs. Further, MC-induced invasion was significantly reduced by anti-CSF-1 treatment while microglia-induced invasion was reduced to a lower extend. Moreover, analysis of lung and breast cancer brain metastasis revealed significant differences of CSF-1 and CSF-1R expression. Taken together, our findings demonstrate not only differences of anti-CSF-1 treatment on MCs and microglia but also in the CSF-1 receptor and ligand expression in brain and bone marrow as well as in brain metastasis.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium pyruvate, Vetec, reagent grade, 98%
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
IL-34 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution