Merck
Evaluation of a porcine model of early aortic valve sclerosis.
Cardiovascular pathology : the official journal of the Society for Cardiovascular Pathology (2014-07-08)
Krista L Sider, Cuilan Zhu, Andrea V Kwong, Zahra Mirzaei, Cornelius F M de Langé, Craig A Simmons
ABSTRACT
Calcific aortic valve disease (CAVD) is associated with significant cardiovascular morbidity. While late-stage CAVD is well-described, early pathobiological processes are poorly understood due to the lack of animal models that faithfully replicate early human disease. Here we evaluated a hypercholesterolemic porcine model of early diet-induced aortic valve sclerosis. Yorkshire swine were fed either a standard or high-fat/high-cholesterol diet for 2 or 5 months. Right coronary aortic valve leaflets were excised and analyzed (immuno)histochemically. Early human-like proteoglycan-rich onlays formed between the endothelial layer and elastic lamina in the fibrosa layer of valve leaflets, with accelerated formation associated with hypercholesterolemia (P<.05). Lipid deposition was more abundant in hypercholesterolemic swine (P<.001), but was present in a minority (28%) of onlays. No myofibroblasts, MAC387-positive macrophages, or fascin-positive dendritic cells were detected in 2-month onlays, with only scarce myofibroblasts present at 5 months. Cells that expressed osteochondral markers Sox9 and Msx2 were preferentially found in dense proteoglycan-rich onlays (P<.05) and with hypercholesterolemia (P<.05). Features of more advanced human CAVD, including calcification, were not observed in this necessarily short study. Early aortic valve sclerosis in hypercholesterolemic swine is characterized by the formation of proteoglycan-rich onlays in the fibrosa, which can occur prior to significant lipid accumulation, inflammatory cell infiltration, or myofibroblast activation. These characteristics mimic those of early human aortic valve disease, and thus the porcine model has utility for the study of early valve sclerosis.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC