Merck
  • MicroRNA-29a suppresses the growth, migration, and invasion of lung adenocarcinoma cells by targeting carcinoembryonic antigen-related cell adhesion molecule 6.
MicroRNA-29a suppresses the growth, migration, and invasion of lung adenocarcinoma cells by targeting carcinoembryonic antigen-related cell adhesion molecule 6.
FEBS letters (2014-08-31)
Hye Sook Han, Seung-Myoung Son, Jieun Yun, Yeong Nang Jo, Ok-Jun Lee
ABSTRACT
Carcinoembryonic antigen-related cell adhesion molecule 6 (CEACAM6) is an important regulator of cell adhesion, invasion, and metastasis. The aim of this study was to evaluate the functional roles of CEACAM6 in lung adenocarcinoma and to identify miRNAs that inhibit the growth, migration, and invasion of lung adenocarcinoma cells by targeting CEACAM6. CEACAM6 expression is associated with poor prognosis of patients with lung adenocarcinoma, and CEACAM6 has important functional roles in controlling the growth, migration, and invasion of lung adenocarcinoma cells in vitro and in vivo. Furthermore, miR-29a can suppress the growth, migration, and invasion of lung adenocarcinoma cells by targeting CEACAM6. Therefore, miR-29a/CEACAM6 axis represents a potential therapeutic target for treatment of lung adenocarcinoma.
Product Description
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%