Merck
  • Beneficial effects of diminished production of hydrogen sulfide or carbon monoxide on hypertension and renal injury induced by NO withdrawal.
Beneficial effects of diminished production of hydrogen sulfide or carbon monoxide on hypertension and renal injury induced by NO withdrawal.
British journal of pharmacology (2014-03-07)
Sebastiaan Wesseling, Joost O Fledderus, Marianne C Verhaar, Jaap A Joles
ABSTRACT
Whether NO, carbon monoxide (CO) and hydrogen sulfide (H2 S) compensate for each other when one or more is depleted is unclear. Inhibiting NOS causes hypertension and kidney injury. Both global depletion of H2 S by cystathionine γ-lyase (CSE) gene deletion and low levels of exogenous H2 S cause hypertension. Inhibiting CO-producing enzyme haeme oxygenase-1 (HO-1) makes rodents hypersensitive to hypertensive stimuli. We hypothesized that combined inhibition of NOS and HO-1 exacerbates hypertension and renal injury, but how combined inhibition of NOS and CSE affect hypertension and renal injury was unclear. Rats were treated with inhibitors of NOS (L-nitroarginine; LNNA), CSE (DL-propargylglycine; PAG), or HO-1 (tin protoporphyrin; SnPP) singly for 1 or 4 weeks or in combinations for 4 weeks. LNNA always reduced NO, decreased H2 S and increased CO after 4 weeks. PAG abolished H2 S, always enhanced CO and reduced NO, but not when used in combination with other inhibitors. SnPP always increased NO, enhanced H2 S and inhibited CO after 1 week. Rats treated with LNNA, but not PAG and SnPP, rapidly developed hypertension followed by renal dysfunction. LNNA-induced hypertension was ameliorated and renal dysfunction prevented by all additional treatments. Renal HO-1 expression was increased by LNNA in injured tubules and increased in all tubules by all other treatments. The amelioration of LNNA-induced hypertension and renal injury by additional inhibition of H2 S and/or CO-producing enzymes appeared to be associated with secondary increases in renal CO or NO production.
Product Description
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
Tin(II) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride, LR, ≥98%
Sigma-Aldrich
Tin(II) chloride, AR, ≥99%
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%
Sigma-Aldrich
Protoporphyrin IX, ≥95%
Sigma-Aldrich
DL-Propargylglycine, cystathionine γ-lyase inhibitor
Sigma-Aldrich
Tin(II) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride, reagent grade, 98%