Merck
  • The mitochondrial permeability transition pore is a dispensable element for mitochondrial calcium efflux.
The mitochondrial permeability transition pore is a dispensable element for mitochondrial calcium efflux.
Cell calcium (2014-04-24)
Elena De Marchi, Massimo Bonora, Carlotta Giorgi, Paolo Pinton
ABSTRACT
The mitochondrial permeability transition pore (mPTP) has long been known to have a role in mitochondrial calcium (Ca(2+)) homeostasis under pathological conditions as a mediator of the mitochondrial permeability transition and the activation of the consequent cell death mechanism. However, its role in the context of mitochondrial Ca(2+) homeostasis is not yet clear. Several studies that were based on PPIF inhibition or knock out suggested that mPTP is involved in the Ca(2+) efflux mechanism, while other observations have revealed the opposite result. The c subunit of the mitochondrial F1/FO ATP synthase has been recently found to be a fundamental component of the mPTP. In this work, we focused on the contribution of the mPTP in the Ca(2+) efflux mechanism by modulating the expression of the c subunit. We observed that forcing mPTP opening or closing did not impair mitochondrial Ca(2+) efflux. Therefore, our results strongly suggest that the mPTP does not participate in mitochondrial Ca(2+) homeostasis in a physiological context in HeLa cells.
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, Vetec, reagent grade, ≥98%, RNase and DNase free
SAFC
BIS-TRIS
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
SAFC
BIS-TRIS
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution