Merck
Expression and prognostic value of Ars2 in hepatocellular carcinoma.
International journal of clinical oncology (2013-11-26)
Qian He, Yongde Huang, Lei Cai, Shaobo Zhang, Chenghua Zhang
ABSTRACT
Hepatocellular carcinoma (HCC) is one of the most common malignant tumors in China. Arsenic resistance protein 2 (Asr2) was reported to be important for microRNA (miR) biogenesis, and its depletion could reduce the levels of several miRs, including miR-21, which is over-expressed in HCC. We hypothesized that Ars2 is also overexpressed in HCC and may be involved in the biological properties of HCC. Ars2 immunolabeling was evaluated in 132 HCCs. Ars2 immunolabeling, Ars2 qRT-PCR and miR-21 were evaluated in 20 HCCs and in paired normal tissues. Ars2 shRNA was transfected into SMCC-7721 and HepG2 HCC cells. The cell proliferation and expression of Ars2 and miR-21 were subsequently evaluated. Ars2 was expressed primarily in the nucleus of HCC cells. The expression of Ars2 was statistically correlated with the loss of HCC differentiation and pathological stage. The survival rates of patients with low Ars2 expression in HCC were statistically higher than patients with overexpressed Ars2 in HCC. Ars2 and miR-21 were more highly expressed in HCC specimens than normal tissues, and they were also correlated. The knockdown of Ars2 in HCC cells inhibited miR-21 expression and cell proliferation. Ars2 is overexpressed in HCC and may have prognostic value; it might play an important role in HCC proliferation and miR-21 expression.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Anti-NS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture