Merck
  • Vitronectin-binding PAI-1 protects against the development of cardiac fibrosis through interaction with fibroblasts.
Vitronectin-binding PAI-1 protects against the development of cardiac fibrosis through interaction with fibroblasts.
Laboratory investigation; a journal of technical methods and pathology (2014-04-02)
Jianyong Zhong, Hai-Chun Yang, Valentina Kon, Agnes B Fogo, Daniel A Lawrence, Ji Ma
ABSTRACT
Plasminogen activator inhibitor-1 (PAI-1) promotes or abates fibrotic processes occurring in different organs. Binding of PAI-1 to vitronectin, an extracellular matrix component, may inhibit vitronectin-integrin complex-mediated cellular responses in pathophysiological conditions. To investigate the importance of plasmin suppression vs vitronectin-binding pathways of PAI-1 in cardiac fibrosis, we studied uninephrectomized mice fed a high salt diet and infused with angiotensin II (Ang II) together with different PAI-1 variants, including PAI-1AK (AK) that inhibits plasminogen activators but does not bind vitronectin, PAI-1RR (RR) that binds vitronectin but does not have protease inhibitory effects or control PAI-1 (CPAI), the control mutant that has similar molecular backbone and half-life as AK and RR while retaining all functions of native PAI-1. Compared with RR and CPAI, non-vitronectin-binding AK significantly increased expression of cardiac fibroblast marker, periostin (Ang+AK 8.40±3.55 vs Ang+RR 2.23±0.44 and Ang+CPAI 2.33±0.12% positive area, both P<0.05) and cardiac fibrosis (Ang+AK 1.79±0.26% vs Ang+RR 0.91±0.18% and Ang+CPAI 0.81±0.12% fibrotic area, both P<0.05), as well as Col1 mRNA (Ang+AK 12.81±1.84 vs Ang+RR 4.04±1.06 and Ang+CPAI 5.23±1.21 fold increase, both P<0.05). To elucidate mechanisms underlying the protective effects of vitronectin-binding PAI-1 against fibrosis, fibroblasts from normal adult human ventricles were stimulated with Ang and different PAI-1 variants. Protease inhibitory AK and CPAI increased supernatant fibronectin, while decreasing plasminogen activator/plasmin activities and matrix metalloproteinase. RR and CPAI variants significantly reduced fibroblast expression of integrin β3, vitronectin level in the supernatant and fibroblast adhesion to vitronectin compared with the non-vitronectin-binding AK. Further, RR and CPAI preserved apoptotic, decreased anti-apoptotic and proliferative activities in fibroblasts. Thus, PAI-1 promotes or protects against development of cardiac fibrosis differentially through the protease inhibitory pathway or through its binding to vitronectin.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Creatinine, anhydrous, ≥98%