Merck
  • cDNA cloning and expression analysis of farnesyl pyrophosphate synthase from Ornithogalum saundersiae.
cDNA cloning and expression analysis of farnesyl pyrophosphate synthase from Ornithogalum saundersiae.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2014-07-30)
Lei Guo, Jian-Qiang Kong
ABSTRACT
Farnesyl pyrophosphate synthase (FPPS, EC 2.5.1.10) catalyzes the consecutive head-to-tail condensations of isopentenyl diphosphate (IPP) with dimethylallyl diphosphate (DMAPP) to form farnesyl pyrophosphate (FPP), a key precursor of sesquiterpenoids, triterpenoids, sterols, and farnesylated proteins. Here we report the molecular cloning and functional identification of a new full-length cDNA encoding FPPS from Ornithogalum saundersiae, a potential medicinal plant that produces a promising antitumour sterol glycoside, OSW-1. An 1327 bp long unigene with an open reading frame of 1044 bp was retrieved from the transcriptome sequencing of O. saundersiae. The full-length FPPS cDNA, designated OsaFPPS, was isolated from O. saundersiae with gene-specific primers. The resultant OsaFPPS encodes a 347-amino acids protein with a calculated molecular mass of 40,085.6 Da, and a theoretical isoelectric point of 5.01. Phylogenetic tree analysis indicated that OsaFPPS belongs to the plant FPPS super-family. Expression of soluble OsaFPPS in E. coli was verified by sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE) and Western blot analysis. Functional analysis of the purified OsaFPPS protein was carried out using IPP and DMAPP as substrates, and the product was unambiguously determined by gas chromatography-mass spectrometry (GC-MS) analyses.
Sigma-Aldrich
γ,γ-Dimethylallyl pyrophosphate triammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≄90% (TLC)