Merck
Online monitoring of myocardial bioprosthesis for cardiac repair.
International journal of cardiology (2014-05-14)
Cristina Prat-Vidal, Carolina Gálvez-Montón, Verónica Puig-Sanvicens, Benjamin Sanchez, Idoia Díaz-Güemes, Paco Bogónez-Franco, Isaac Perea-Gil, Anna Casas-Solà, Santiago Roura, Aida Llucià-Valldeperas, Carolina Soler-Botija, Francisco M Sánchez-Margallo, Carlos E Semino, Ramon Bragos, Antoni Bayes-Genis
ABSTRACT
The aim of this study was to develop a myocardial bioprosthesis for cardiac repair with an integrated online monitoring system. Myocardial infarction (MI) causes irreversible myocyte loss and scar formation. Tissue engineering to reduce myocardial scar size has been tested with variable success, yet scar formation and modulation by an engineered graft is incompletely characterized. Decellularized human pericardium was embedded using self-assembling peptide RAD16-I with or without GFP-labeled mediastinal adipose tissue-derived progenitor cells (MATPCs). Resulting bioprostheses were implanted over the ischemic myocardium in the swine model of MI (n=8 treated and n=5 control animals). For in vivo electrical impedance spectroscopy (EIS) monitoring, two electrodes were anchored to construct edges, covered by NanoGold particles and connected to an impedance-based implantable device. Histological evaluation was performed to identify and characterize GFP cells on post mortem myocardial sections. Pluripotency, cardiomyogenic and endothelial potential and migratory capacity of porcine-derived MATPCs were demonstrated in vitro. Decellularization protocol efficiency, biodegradability, as well as in vitro biocompatibility after recellularization were also verified. One month after myocardial bioprosthesis implantation, morphometry revealed a 36% reduction in infarct area, Ki67(+)-GFP(+)-MATPCs were found at infarct core and border zones, and bioprosthesis vascularization was confirmed by presence of Griffonia simplicifolia lectin I (GSLI) B4 isolectin(+)-GFP(+)-MATPCs. Electrical impedance measurement at low and high frequencies (10 kHz-100 kHz) allowed online monitoring of scar maturation. With clinical translation as ultimate goal, this myocardial bioprosthesis holds promise to be a viable candidate for human cardiac repair.
Sigma-Aldrich
Sodium dodecyl sulfate, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O