Merck
  • A Novel Human Radixin Peptide Inhibits Hepatitis C Virus Infection at the Level of Cell Entry.
A Novel Human Radixin Peptide Inhibits Hepatitis C Virus Infection at the Level of Cell Entry.
International journal of peptide research and therapeutics (2014-11-08)
Terence N Bukong, Karen Kodys, Gyongyi Szabo
ABSTRACT
Hepatitis C virus infection of hepatocytes is a multistep process involving the interaction between viral and host cell molecules. Recently, we identified ezrin-moesin-radixin proteins and spleen tyrosine kinase (SYK) as important host therapeutic targets for HCV treatment development. Previously, an ezrin hinge region peptide (Hep1) has been shown to exert anti-HCV properties in vivo, though its mechanism of action remains limited. In search of potential novel inhibitors of HCV infection and their functional mechanism we analyzed the anti-HCV properties of different human derived radixin peptides. Sixteen different radixin peptides were derived, synthesized and tested. Real-time quantitative PCR, cell toxicity assay, immuno-precipitation/western blot analysis and computational resource for drug discovery software were used for experimental analysis. We found that a human radixin hinge region peptide (Peptide1) can specifically block HCV J6/JFH-1 infection of Huh7.5 cells. Peptide 1 had no cell toxicity or intracellular uptake into Huh7.5 cells. Mechanistically, the anti-HCV activity of Peptide 1 extended to disruption of HCV engagement of CD81 thereby blocking downstream SYK activation, which we have recently demonstrated to be important for effective HCV infection of target hepatocytes. Our findings highlight a novel functional class of anti-HCV agents that can inhibit HCV infection, most likely by disrupting vital viral-host signaling interactions at the level of virus entry.
Sigma-Aldrich
Anti-NS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, LR, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, AR, ≥99.5%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial