Merck
  • dCK expression correlates with 5-fluorouracil efficacy and HuR cytoplasmic expression in pancreatic cancer: a dual-institutional follow-up with the RTOG 9704 trial.
dCK expression correlates with 5-fluorouracil efficacy and HuR cytoplasmic expression in pancreatic cancer: a dual-institutional follow-up with the RTOG 9704 trial.
Cancer biology & therapy (2014-03-13)
Florencia McAllister, Danielle M Pineda, Masaya Jimbo, Shruti Lal, Richard A Burkhart, Jennifer Moughan, Kathryn A Winter, Kotb Abdelmohsen, Myriam Gorospe, Ana de Jesus Acosta, Rachana H Lankapalli, Jordan M Winter, Charles J Yeo, Agnieska K Witkiewicz, Christine A Iacobuzio-Donahue, Daniel Laheru, Jonathan R Brody
ABSTRACT
Deoxycytidine kinase (dCK) and human antigen R (HuR) have been associated with response to gemcitabine in small studies. The present study investigates the prognostic and predictive value of dCK and HuR expression levels for sensitivity to gemcitabine and 5-fluorouracil (5-FU) in a large phase III adjuvant trial with chemoradiation backbone in pancreatic ductal adenocarcinoma (PDA). The dCK and HuR expression levels were determined by immunohistochemistry on a tissue microarray of 165 resected PDAs from the Radiation Therapy Oncology Group (RTOG) 9704 trial. Association with overall survival (OS) and disease-free survival (DFS) status were analyzed using the log-rank test and the Cox proportional hazards model. Experiments with cultured PDA cells were performed to explore mechanisms linking dCK and HuR expression to drug sensitivity. dCK expression levels were associated with improved OS for all patients analyzed from RTOG 9704 (HR: 0.66, 95% CI [0.47-0.93], P = 0.015). In a subset analysis based on treatment arm, the effect was restricted to patients receiving 5-FU (HR: 0.53, 95% CI [0.33-0.85], P = 0.0078). Studies in cultured cells confirmed that dCK expression rendered cells more sensitive to 5-FU. HuR cytoplasmic expression was neither prognostic nor predictive of treatment response. Previous studies along with drug sensitivity and biochemical studies demonstrate that radiation interferes with HuR's regulatory effects on dCK, and could account for the negative findings herein based on the clinical study design (i.e., inclusion of radiation). Finally, we demonstrate that 5-FU can increase HuR function by enhancing HuR translocation from the nucleus to the cytoplasm, similar to the effect of gemcitabine in PDA cells. For the first time, in the pre-treatment tumor samples, dCK and HuR cytoplasmic expression were strongly correlated (chi-square P = 0.015). This dual-institutional follow up study, in a multi-institutional PDA randomized clinical trial, observed that dCK expression levels were prognostic and had predictive value for sensitivity to 5-FU.
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)