Merck
  • Instability of trinucleotidic repeats during chromatin remodeling in spermatids.
Instability of trinucleotidic repeats during chromatin remodeling in spermatids.
Human mutation (2014-08-20)
Olivier Simard, Marie-Chantal Grégoire, Mélina Arguin, Marc-André Brazeau, Frédéric Leduc, Isabelle Marois, Martin V Richter, Guylain Boissonneault
ABSTRACT
Transient DNA breaks and evidence of DNA damage response have recently been reported during the chromatin remodeling process in haploid spermatids, creating a potential window of enhanced genetic instability. We used flow cytometry to achieve separation of differentiating spermatids into four highly purified populations using transgenic mice harboring 160 CAG repeats within exon 1 of the human Huntington disease gene (HTT). Trinucleotic repeat expansion was found to occur immediately following the chromatin remodeling steps, confirming the genetic instability of the process and pointing to the origin of paternal anticipation observed in some trinucleotidic repeats diseases.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea, AR, ≥99%
Sigma-Aldrich
Urea, LR, ≥99%
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, Vetec, reagent grade, 99%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, 99% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, meets USP testing specifications
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)