Merck
  • Immunoregulatory function of PIR-A/B+ DCs in the inflammatory responses of dextran sodium sulfate-induced colitis.
Immunoregulatory function of PIR-A/B+ DCs in the inflammatory responses of dextran sodium sulfate-induced colitis.
Journal of gastroenterology (2013-10-01)
Akiko Kurishima, Muneo Inaba, Yutaku Sakaguchi, Toshiro Fukui, Kazushige Uchida, Akiyoshi Nishio, Shosaku Nomura, Kazuichi Okazaki
ABSTRACT
Dendritic cells (DCs) may play an important role in forms of inflammatory bowel disease (IBD), such as Crohn's disease and ulcerative colitis. DCs are generally recognized as initiators of acquired immunity and also serve as regulators of both innate and acquired immunity. We used the animal model of colitis induced by dextran sodium sulfate (DSS), and examined whether DCs prepared from the colon show immunoregulatory roles in the termination of DSS-induced colitis. C57BL/6 mice exposed to DSS for 5 days developed acute colitis. DCs were isolated from the large intestinal lamina propria, and then analyzed for phenotypical, functional, and genetic data. Only PIR-A/B(low) conventional DCs (cDCs) were detected in the steady state. However, after the treatment of DSS, PIR-A/B(high) cDCs appeared and gradually increased from day 5 to day 7, at which time the DSS-induced colitis was terminated. Then, allogeneic mixed leukocyte reaction (MLR) was performed. The stimulatory activity of PIR-A/B(high) cDCs obtained on day 7 was very low, and the addition of PIR-A/B(high) cDCs suppressed the T cell proliferation in MLR, indicating the immunoregulatory role of PIR-A/B(high) cDCs. The immunoregulatory role of PIR-A/B(high) cDCs was confirmed by the in vivo transfer experiment, showing their therapeutic effect on DSS-induced colitis. The message level of TGFβi was significantly higher in PIR-A/B(high) cDCs, while that of IFN-γ was highly upregulated in PIR-A/B(low) cDCs, being well in accordance with the fact that PIR-A/B(high) cDCs showed a suppressive function against activated T cells. PIR-A/B(high) cDCs showed a suppressive function against activated T cells by producing inhibitory cytokines.
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0% (RT), BioUltra
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%