Merck
  • Immunohistochemical characteristics of regenerated bone after surgical therapy of advanced ligature-induced peri-implantitis defects.
Immunohistochemical characteristics of regenerated bone after surgical therapy of advanced ligature-induced peri-implantitis defects.
Clinical oral investigations (2013-11-26)
Frank Schwarz, Ilja Mihatovic, Vladimir Golubovic, Jürgen Becker, Martin Sager
ABSTRACT
This study seeks to assess and compare immunohistochemical characteristics of regenerated and pristine bone areas following surgical therapy of advanced peri-implantitis. At ligature-induced peri-implantitis defects, the intrabony component was filled with a natural bone mineral (NBM), and the supracrestal component was treated by either an equine bone block (EB) or implantoplasty. NBM and EB were soak-loaded with rhBMP-2 or sterile saline. Membrane (i.e., native collagen) protected sites were submerged for 12 weeks. Osteocalcin (OC) and transglutaminase 2 (TG2; angiogenesis) antigen reactivity was assessed within the augmented-(AA) and pristine bone (PB) areas at non-exposed sites (n = 39 defects). In all groups investigated, mean OC (AA, 0.5 ± 0.4 to 1.9 ± 2.9 %/PB, 1.7 ± 2.6 to 3.5 ± 6.5 %) and TG2 (AA, 0.6 ± 0.5 to 1.3 ± 1.5 %/PB, 0.5 ± 0.5 to 1.6 ± 1.9 %) values within AA did not significantly differ from those values assessed within PB (P > 0.05, respectively). AA formed in different treatment groups may not be considered as qualitatively (i.e., OC and TG2) compromised bone.
Product Description
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, ≥30%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, LR, contains sodium pyrophosphate as stabilizer, ≥30 % (w/v)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, LR, contains sodium pyrophosphate as stabilizer, ≥50 % (w/v)
Sigma-Aldrich
Hydrogen peroxide solution, AR, contains sodium pyrophosphate as stabilizer, ≥30 % (w/v)
Sigma-Aldrich
Hydrogen peroxide solution, ≥30%