Merck
  • High prognostic value of minimal residual disease detected by flow-cytometry-enhanced fluorescence in situ hybridization in core-binding factor acute myeloid leukemia (CBF-AML).
High prognostic value of minimal residual disease detected by flow-cytometry-enhanced fluorescence in situ hybridization in core-binding factor acute myeloid leukemia (CBF-AML).
Annals of hematology (2014-05-23)
Libing Wang, Lei Gao, Sheng Xu, Shenglan Gong, Min Liu, Huiying Qiu, Xiaoqian Xu, Xiong Ni, Li Chen, Shuqing Lu, Jie Chen, Xianmin Song, Weiping Zhang, Jianmin Yang, Xiaoxia Hu, Jianmin Wang
ABSTRACT
Acute myeloid leukemia (AML) is generally regarded as a disorder of stem cells, known as leukemic initiating cells (LICs), which initiate the disease and contribute to relapses. Although the phenotype of these cells remains unclear in most patients, they are enriched within the CD34(+)CD38(-) population. In core-binding factor (CBF) AML, the cytogenetic abnormalities also exist in LIC. The aim of this study was to determine the prognostic power of minimal residual disease (MRD) measured by fluorescence in situ hybridization (FISH) in CD34(+)CD38(-) cells sorted by flow cytometry at different periods during therapy. Thirty-six patients under 65 years of age with de novo CBF-AML treated with intensive chemotherapy were retrospectively included in this study. Correlations with relapse-free survival (RFS) and overall survival were evaluated with univariate and multivariate analyses. FISH efficiently identified LICs in the CD34(+)CD38(-) population. The presence of FISH(+)CD34(+)CD38(-) cells before consolidation was negatively associated with cumulative incidence of relapse (64 vs 18 %, P = .012), which showed prognostic value for RFS (12 vs 68 %, P = .008) and OS (11 vs 75 %, P = .0005), and retained prognostic significance for RFS in multivariate analysis. The detection of FISH(+)CD34(+)CD38(-) cells before consolidation therapy significantly correlated with long-term survival. Fluorescence-activated cell sorting (FACS)-FISH could be potentially adopted as a MRD monitor approach in clinical practice to identify CBF-AML patients at risk of treatment failure during therapy.
Product Description
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, LR, ≥99.5%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, AR, ≥99.8%
Sigma-Aldrich
Acetic acid, suitable for HPLC, ≥99.8%
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%