Merck
  • Plasmacytoid dendritic cells support melanoma progression by promoting Th2 and regulatory immunity through OX40L and ICOSL.
Plasmacytoid dendritic cells support melanoma progression by promoting Th2 and regulatory immunity through OX40L and ICOSL.
Cancer immunology research (2014-04-30)
Caroline Aspord, Marie-Therese Leccia, Julie Charles, Joel Plumas
ABSTRACT
Even though melanoma is considered to be one of the most immunogenic solid tumors, handling its development remains a challenge. The basis for such escape from antitumor immune control has not yet been documented. Plasmacytoid dendritic cells (pDC) are emerging as crucial but still enigmatic cells in cancer. In melanoma, the function of tumor-infiltrating pDCs remains poorly explored. We investigated the pathophysiologic role of pDCs in melanoma, both ex vivo from a large cohort of melanoma patients and in vivo in melanoma-bearing humanized mice. pDCs were found in high proportions in cutaneous melanoma and tumor-draining lymph nodes, yet associated with poor clinical outcome. We showed that pDCs migrating to the tumor microenvironment displayed particular features, subsequently promoting proinflammatory Th2 and regulatory immune profiles through OX40L and ICOSL expression. Elevated frequencies of interleukin (IL)-5-, IL-13- and IL-10-producing T cells in patients with melanoma correlated with high proportions of OX40L- and ICOSL-expressing pDCs. Strikingly TARC/CCL17, MDC/CCL22, and MMP-2 found in the melanoma microenvironment were associated with pDC accumulation, OX40L and ICOSL modulation, and/or early relapse. Thus, melanoma actively exploits pDC plasticity to promote its progression. By identifying novel insights into the mechanism of hijacking of immunity by melanoma, our study exposes potential for new therapeutic opportunities.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%