Merck
  • Vectored antibody gene delivery protects against Plasmodium falciparum sporozoite challenge in mice.
Vectored antibody gene delivery protects against Plasmodium falciparum sporozoite challenge in mice.
Proceedings of the National Academy of Sciences of the United States of America (2014-08-13)
Cailin Deal, Alejandro B Balazs, Diego A Espinosa, Fidel Zavala, David Baltimore, Gary Ketner
ABSTRACT
Malaria caused by Plasmodium falciparum kills nearly one million children each year and imposes crippling economic burdens on families and nations worldwide. No licensed vaccine exists, but infection can be prevented by antibodies against the circumsporozoite protein (CSP), the major surface protein of sporozoites, the form of the parasite injected by mosquitoes. We have used vectored immunoprophylaxis (VIP), an adeno-associated virus-based technology, to introduce preformed antibody genes encoding anti-P. falciparum CSP mAb into mice. VIP vector-transduced mice exhibited long-lived mAb expression at up to 1,200 µg/mL in serum, and up to 70% were protected from both i.v. and mosquito bite challenge with transgenic Plasmodium berghei rodent sporozoites that incorporate the P. falciparum target of the mAb in their CSP. Serum antibody levels and protection from mosquito bite challenge were dependent on the dose of the VIP vector. All individual mice expressing CSP-specific mAb 2A10 at 1 mg/mL or more were completely protected, suggesting that in this model system, exceeding that threshold results in consistent sterile protection. Our results demonstrate the potential of VIP as a path toward the elusive goal of immunization against malaria.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, low water for titration
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, AR, ≥99.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, LR, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard