Merck
  • Evaluation of the mutagenicity and genotoxic potential of carvacrol and thymol using the Ames Salmonella test and alkaline, Endo III- and FPG-modified comet assays with the human cell line Caco-2.
Evaluation of the mutagenicity and genotoxic potential of carvacrol and thymol using the Ames Salmonella test and alkaline, Endo III- and FPG-modified comet assays with the human cell line Caco-2.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-07-20)
Maria LLana-Ruiz-Cabello, Sara Maisanaba, Maria Puerto, Ana I Prieto, Silvia Pichardo, Ángeles Jos, Ana M Cameán
ABSTRACT
Currently, direct antimicrobial and antioxidant additives derived from essential oils are used in food packaging and are perceived by consumers as low-health-risk compounds. In this study, we investigated the potential mutagenicity and genotoxicity of carvacrol and thymol, major compounds in several essential oils, using the Ames Salmonella test and the alkaline, Endo III- and formamidopyrimidine glycosylase (FPG)-modified comet assays, respectively. Thymol did not show any mutagenic activity at any concentration assayed (0-250 μM), whereas carvacrol exhibited mutagenic potential, displaying greater activity in presence of the metabolic fraction (29-460 μM). The genotoxic effects were evaluated in the human colon carcinoma cell line Caco-2, and the standard comet assay revealed that neither carvacrol (0-460 μM) nor thymol (0-250 μM) had any affects at 24 and 48 h. The FPG-modified comet assay showed that the highest concentration of carvacrol (460 μM) caused DNA damage, indicating damage to the purine bases. These results should be used to identify the appropriate concentrations of carvacrol and thymol as additives in food packaging. Moreover, further studies are necessary to explore the safety and/or the toxicity mechanisms of these compounds.
Product Description
Supelco
Thymol, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Thymol, FCC, FG
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Thymol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thymol, ≥98.5%
Supelco
Thymol, analytical standard
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%