Merck
  • Facile and rapid generation of large-scale microcollagen gel array for long-term single-cell 3D culture and cell proliferation heterogeneity analysis.
Facile and rapid generation of large-scale microcollagen gel array for long-term single-cell 3D culture and cell proliferation heterogeneity analysis.
Analytical chemistry (2014-02-11)
Zhichao Guan, Shasha Jia, Zhi Zhu, Mingxia Zhang, Chaoyong James Yang
ABSTRACT
Microfabricated devices are suitable for single-cell analysis due to their high throughput, compatible dimensions and controllable microenvironment. However, existing devices for single-cell culture and analysis encounter some limitations, such as nutrient depletion, random cell migration and complicated fluid shear influence. Moreover, most of the single-cell culture and analysis devices are based on 2D cell culture conditions, even though 3D cell culture methods have been demonstrated to better mimic the real cell microenvironment in vivo. To solve these problems, herein we develop a microcollagen gel array (μCGA) based approach for high-throughput long-term single-cell culture and single-cell analysis under 3D culture conditions. Type-I collagen, a well-established 3D cell culture medium, was used as the scaffold for 3D cell growth. A 2 × 2 cm PDMS chip with 10 000 μCGA units was fabricated to encapsulate thousands of single cells in less than 15 min. Single cells were able to be confined and survive in μCGA units for more than 1 month. The capability of large-scale and long-term single-cell 3D culture under open culture conditions allows us to study cellular proliferation heterogeneity and drug cytotoxicity at the single-cell level. Compared with existing devices for single-cell analysis, μCGA solves the problems of nutrient depletion and random cellular migration, avoids the influence of complicated fluid shear, and mimics the real 3D growth environment in vivo, thereby providing a feasible 3D long-term single-cell culture method for single-cell analysis and drug screening.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered