Merck
  • Fluorescent derivatization method of proteins for characterization by capillary electrophoresis-sodium dodecyl sulfate with laser-induced fluorescence detection.
Fluorescent derivatization method of proteins for characterization by capillary electrophoresis-sodium dodecyl sulfate with laser-induced fluorescence detection.
Analytical chemistry (2007-06-27)
David A Michels, Lowell J Brady, Amy Guo, Alain Balland
ABSTRACT
A fast and improved sample preparation scheme was developed for protein analysis using capillary electrophoresis-sodium dodecyl sulfate (CE-SDS) with laser-induced fluorescence detection. This CE-SDS method was developed as a purity assay for recombinant monoclonal antibodies (rMAbs). In this assay, rMAbs are derivatized with the fluorogenic reagent 3-(2-furoyl)-quinoline-2-carboxaldehyde (FQ) in the presence of a nucleophile (CN-), which fluoresces only upon covalent binding to the protein. Purification after labeling is therefore not necessary to remove unreacted reagents. Proteins are incubated at 75 degrees C for 5 min to facilitate denaturation and labeling. For nonreduced preparation, rMAbs are labeled at pH 6.5 with a dye-to-protein (D/P) molar ratio of 50:1, which forms conjugates having 6 +/- 4 FQ labels. For reduced preparation, rMAbs are labeled at pH 9.3 with a D/P molar ratio of 10:1, which generates light chain conjugates incorporated with 3 +/- 2 FQ labels. Labeling artifacts such as fragmentation or aggregation are absent with use of alkylation reagents. This efficient labeling scheme generates detection limits for FQ-labeled rMAbs as low as 10 ng/mL. In comparison to other labeling strategies, labeling proteins with FQ has the advantage of speed, ease of use, and robust quantification.
Sigma-Aldrich
3-(2-Furoyl)quinoline-2-carboxaldehyde, ≥95% (HPLC), suitable for fluorescence, BioReagent