Merck
  • Synthesis of 1,4-dichloro-1,3-butadienes by rhodium complex catalyzed reaction of terminal alkynes with trichloroacetyl chloride.
Synthesis of 1,4-dichloro-1,3-butadienes by rhodium complex catalyzed reaction of terminal alkynes with trichloroacetyl chloride.
The Journal of organic chemistry (2009-11-20)
Taigo Kashiwabara, Kouichirou Fuse, Takeshi Muramatsu, Masato Tanaka
ABSTRACT
Chlorinative dimerization of terminal alkynes with trichloroacetyl chloride as chlorine donor proceeds in the presence of rhodium catalysts to give (Z,Z)-1,4-dichloro-1,3-butadienes stereoselectively. Ligand screening has revealed that reactions using sterically bulky and electron-donating ligands like trimesitylphosphine are high yielding. The reaction is compatible with a range of functional groups to give the title compounds nearly quantitatively in most cases. A mechanistic possibility involving coupling of beta-chloroalken-1-yl intermediate has been discussed.
Product Description
Supelco
Trichloroacetyl chloride, for GC derivatization, ≥98.5%
Sigma-Aldrich
Trichloroacetyl chloride, 99%