Merck
  • Lethal effect of a single dose of rasburicase in a preterm newborn infant.
Lethal effect of a single dose of rasburicase in a preterm newborn infant.
Pediatrics (2012-12-05)
Patrizia Zaramella, Alessandra De Salvia, Martina Zaninotto, Maura Baraldi, Giovanni Capovilla, Domenico De Leo, Lino Chiandetti
ABSTRACT
This case report describes a preterm newborn infant who was treated with a single dose of rasburicase for an increase in uric acid level. He died on the third day as a result of complications of hemolysis, which appeared to be precipitated by rasburicase. The patient's death was preceded by progressive respiratory insufficiency, lactic acidosis, and hyperbilirubinemia, culminating in refractory hypoxia and hypotension. A postmortem assay for glucose-6-phosphate dehydrogenase showed deficiency and the glucose-6-phosphate dehydrogenase Mediterranean genotype.
Sigma-Aldrich
Uricase from Bacillus fastidiosus, lyophilized, ~9 U/mg
Sigma-Aldrich
Uricase from Candida sp., recombinant, expressed in E. coli, lyophilized powder, ≥2 units/mg solid
Sigma-Aldrich
Uricase from Arthrobacter globiformis, lyophilized powder, 15-30 units/mg protein (biuret)