Merck
  • 3,5-Diiodo-L-thyronine prevents high-fat-diet-induced insulin resistance in rat skeletal muscle through metabolic and structural adaptations.
3,5-Diiodo-L-thyronine prevents high-fat-diet-induced insulin resistance in rat skeletal muscle through metabolic and structural adaptations.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2011-06-15)
Maria Moreno, Elena Silvestri, Rita De Matteis, Pieter de Lange, Assunta Lombardi, Daniela Glinni, Rosalba Senese, Federica Cioffi, Anna Maria Salzano, Andrea Scaloni, Antonia Lanni, Fernando Goglia
ABSTRACT
The worldwide prevalence of obesity-associated pathologies, including type 2 diabetes, requires thorough investigation of mechanisms and interventions. Recent studies have highlighted thyroid hormone analogs and derivatives as potential agents able to counteract such pathologies. In this study, in rats receiving a high-fat diet (HFD), we analyzed the effects of a 4-wk daily administration of a naturally occurring iodothyronine, 3,5-diiodo-L-thyronine (T2), on the gastrocnemius muscle metabolic/structural phenotype and insulin signaling. The HFD-induced increases in muscle levels of fatty acid translocase (3-fold; P<0.05) and TGs (2-fold, P<0.05) were prevented by T2 (each; P<0.05 vs. HFD). T2 increased insulin-stimulated Akt phosphorylation levels (∼2.5-fold; P<0.05 vs. HFD). T2 induced these effects while sparing muscle mass and without cardiac hypertrophy. T2 increased the muscle contents of fast/glycolytic fibers (2-fold; P<0.05 vs. HFD) and sarcolemmal glucose transporter 4 (3-fold; P<0.05 vs. HFD). Adipocyte differentiation-related protein was predominantly present within the slow/oxidative fibers in HFD-T2. In T2-treated rats (vs. HFD), glycolytic enzymes and associated components were up-regulated (proteomic analysis, significance limit: 2-fold; P<0.05), as was phosphofructokinase activity (by 1.3-fold; P<0.05), supporting the metabolic shift toward a more glycolytic phenotype. These results highlight T2 as a potential therapeutic approach to the treatment of diet-induced metabolic dysfunctions.
Sigma-Aldrich
3,5-Diiodo-L-thyronine, thyroid hormone analog