Merck
  • Nsp1 protein of SARS-CoV-2 disrupts the mRNA export machinery to inhibit host gene expression.
Nsp1 protein of SARS-CoV-2 disrupts the mRNA export machinery to inhibit host gene expression.
Science advances (2021-02-07)
Ke Zhang, Lisa Miorin, Tadashi Makio, Ishmael Dehghan, Shengyan Gao, Yihu Xie, Hualin Zhong, Matthew Esparza, Thomas Kehrer, Anil Kumar, Tom C Hobman, Christopher Ptak, Boning Gao, John D Minna, Zhijian Chen, Adolfo García-Sastre, Yi Ren, Richard W Wozniak, Beatriz M A Fontoura
ABSTRACT
The ongoing unprecedented severe acute respiratory syndrome caused by the SARS-CoV-2 outbreak worldwide has highlighted the need for understanding viral-host interactions involved in mechanisms of virulence. Here, we show that the virulence factor Nsp1 protein of SARS-CoV-2 interacts with the host messenger RNA (mRNA) export receptor heterodimer NXF1-NXT1, which is responsible for nuclear export of cellular mRNAs. Nsp1 prevents proper binding of NXF1 to mRNA export adaptors and NXF1 docking at the nuclear pore complex. As a result, a significant number of cellular mRNAs are retained in the nucleus during infection. Increased levels of NXF1 rescues the Nsp1-mediated mRNA export block and inhibits SARS-CoV-2 infection. Thus, antagonizing the Nsp1 inhibitory function on mRNA export may represent a strategy to restoring proper antiviral host gene expression in infected cells.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
ANTI-BAT1(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution