Merck
  • A unique subset of glycolytic tumour-propagating cells drives squamous cell carcinoma.
A unique subset of glycolytic tumour-propagating cells drives squamous cell carcinoma.
Nature metabolism (2021-02-24)
Jee-Eun Choi, Carlos Sebastian, Christina M Ferrer, Caroline A Lewis, Moshe Sade-Feldman, Thomas LaSalle, Anna Gonye, Begona G C Lopez, Walid M Abdelmoula, Michael S Regan, Murat Cetinbas, Gloria Pascual, Gregory R Wojtkiewicz, Giorgia G Silveira, Ruben Boon, Kenneth N Ross, Itay Tirosh, Srinivas V Saladi, Leif W Ellisen, Ruslan I Sadreyev, Salvador Aznar Benitah, Nathalie Y R Agar, Nir Hacohen, Raul Mostoslavsky
ABSTRACT
Head and neck squamous cell carcinoma (SCC) remains among the most aggressive human cancers. Tumour progression and aggressiveness in SCC are largely driven by tumour-propagating cells (TPCs). Aerobic glycolysis, also known as the Warburg effect, is a characteristic of many cancers; however, whether this adaptation is functionally important in SCC, and at which stage, remains poorly understood. Here, we show that the NAD+-dependent histone deacetylase sirtuin 6 is a robust tumour suppressor in SCC, acting as a modulator of glycolysis in these tumours. Remarkably, rather than a late adaptation, we find enhanced glycolysis specifically in TPCs. More importantly, using single-cell RNA sequencing of TPCs, we identify a subset of TPCs with higher glycolysis and enhanced pentose phosphate pathway and glutathione metabolism, characteristics that are strongly associated with a better antioxidant response. Together, our studies uncover enhanced glycolysis as a main driver in SCC, and, more importantly, identify a subset of TPCs as the cell of origin for the Warburg effect, defining metabolism as a key feature of intra-tumour heterogeneity.
Sigma-Aldrich
(S)-(−)-N-(Trifluoroacetyl)pyrrolidine-2-carbonyl chloride solution, 0.1 M in methylene chloride
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-MPC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Propidium iodide solution