Merck
  • Rad54 Drives ATP Hydrolysis-Dependent DNA Sequence Alignment during Homologous Recombination.
Rad54 Drives ATP Hydrolysis-Dependent DNA Sequence Alignment during Homologous Recombination.
Cell (2020-06-06)
J Brooks Crickard, Corentin J Moevus, Youngho Kwon, Patrick Sung, Eric C Greene
ABSTRACT
Homologous recombination (HR) helps maintain genome integrity, and HR defects give rise to disease, especially cancer. During HR, damaged DNA must be aligned with an undamaged template through a process referred to as the homology search. Despite decades of study, key aspects of this search remain undefined. Here, we use single-molecule imaging to demonstrate that Rad54, a conserved Snf2-like protein found in all eukaryotes, switches the search from the diffusion-based pathways characteristic of the basal HR machinery to an active process in which DNA sequences are aligned via an ATP-dependent molecular motor-driven mechanism. We further demonstrate that Rad54 disrupts the donor template strands, enabling the search to take place within a migrating DNA bubble-like structure that is bound by replication protein A (RPA). Our results reveal that Rad54, working together with RPA, fundamentally alters how DNA sequences are aligned during HR.
Sigma-Aldrich
SUMO Protease, His tagged recombinant protein, lyophilized powder
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%