Merck
  • Rab27a co-ordinates actin-dependent transport by controlling organelle-associated motors and track assembly proteins.
Rab27a co-ordinates actin-dependent transport by controlling organelle-associated motors and track assembly proteins.
Nature communications (2020-07-15)
Noura Alzahofi, Tobias Welz, Christopher L Robinson, Emma L Page, Deborah A Briggs, Amy K Stainthorp, James Reekes, David A Elbe, Felix Straub, Wouter W Kallemeijn, Edward W Tate, Philip S Goff, Elena V Sviderskaya, Marta Cantero, Lluis Montoliu, Francois Nedelec, Amanda K Miles, Maryse Bailly, Eugen Kerkhoff, Alistair N Hume
ABSTRACT
Cell biologists generally consider that microtubules and actin play complementary roles in long- and short-distance transport in animal cells. On the contrary, using melanosomes of melanocytes as a model, we recently discovered that the motor protein myosin-Va works with dynamic actin tracks to drive long-range organelle dispersion in opposition to microtubules. This suggests that in animals, as in yeast and plants, myosin/actin can drive long-range transport. Here, we show that the SPIRE-type actin nucleators (predominantly SPIRE1) are Rab27a effectors that co-operate with formin-1 to generate actin tracks required for myosin-Va-dependent transport in melanocytes. Thus, in addition to melanophilin/myosin-Va, Rab27a can recruit SPIREs to melanosomes, thereby integrating motor and track assembly activity at the organelle membrane. Based on this, we suggest a model in which organelles and force generators (motors and track assemblers) are linked, forming an organelle-based, cell-wide network that allows their collective activity to rapidly disperse the population of organelles long-distance throughout the cytoplasm.
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Triethylammonium bicarbonate buffer, 1.0 M, pH 8.5±0.1
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99% (titration)