Merck
  • Depletion of the AD Risk Gene SORL1 Selectively Impairs Neuronal Endosomal Traffic Independent of Amyloidogenic APP Processing.
Depletion of the AD Risk Gene SORL1 Selectively Impairs Neuronal Endosomal Traffic Independent of Amyloidogenic APP Processing.
Cell reports (2020-06-04)
Allison Knupp, Swati Mishra, Refugio Martinez, Jacquelyn E Braggin, Marcell Szabo, Chizuru Kinoshita, Dale W Hailey, Scott A Small, Suman Jayadev, Jessica E Young
ABSTRACT
SORL1/SORLA is a sorting receptor involved in retromer-related endosomal traffic and an Alzheimer's disease (AD) risk gene. Using CRISPR-Cas9, we deplete SORL1 in hiPSCs to ask if loss of SORL1 contributes to AD pathogenesis by endosome dysfunction. SORL1-deficient hiPSC neurons show early endosome enlargement, a hallmark cytopathology of AD. There is no effect of SORL1 depletion on endosome size in hiPSC microglia, suggesting a selective effect on neuronal endosomal trafficking. We validate defects in neuronal endosomal traffic by showing altered localization of amyloid precursor protein (APP) in early endosomes, a site of APP cleavage by the β-secretase (BACE). Inhibition of BACE does not rescue endosome enlargement in SORL1-deficient neurons, suggesting that this phenotype is independent of amyloidogenic APP processing. Our data, together with recent findings, underscore how sporadic AD pathways regulating endosomal trafficking and autosomal-dominant AD pathways regulating APP cleavage independently converge on the defining cytopathology of AD.
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
LY2886721, ≥98% (HPLC)
Sigma-Aldrich
1-Thioglycerol, ≥97%
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Triton X-100, laboratory grade