Merck
Bioorthogonal Chemical Activation of Kinases in Living Systems.
ACS central science (2016-06-10)
Gong Zhang, Jie Li, Ran Xie, Xinyuan Fan, Yanjun Liu, Siqi Zheng, Yun Ge, Peng R Chen
ABSTRACT
Selective manipulation of protein kinases under living conditions is highly desirable yet extremely challenging, particularly in a gain-of-function fashion. Here we employ our recently developed bioorthogonal cleavage reaction as a general strategy for intracellular activation of individual kinases. Site-specific incorporation of trans-cyclooctene-caged lysine in place of the conserved catalytic lysine, in conjunction with the cleavage partner dimethyl-tetrazine, allowed efficient lysine decaging with the kinase activity chemically rescued in living systems.
Sigma-Aldrich
N6-[[(1α,8α,9α)-Bicyclo[6.1.0]non-4-yn-9-ylmethoxy]carbonyl]-L-lysine, ≥95%
Sigma-Aldrich
Methyl-o-nitropiperonyllysine, ≥95%