Merck
  • High glucose reduces expression of podocin in cultured human podocytes by stimulating TRPC6.
High glucose reduces expression of podocin in cultured human podocytes by stimulating TRPC6.
American journal of physiology. Renal physiology (2019-10-01)
Xiao-Yu Lu, Bing-Chen Liu, Yu-Ze Cao, Chang Song, Hua Su, Guangping Chen, Janet D Klein, Hui-Xue Zhang, Li-Hua Wang, He-Ping Ma
ABSTRACT
The transient receptor potential canonical 6 (TRPC6) channel and podocin are colocalized in the glomerular slit diaphragm as an important complex to maintain podocyte function. Gain of TRPC6 function and loss of podocin function induce podocyte injury. We have previously shown that high glucose induces apoptosis of podocytes by activating TRPC6; however, whether the activated TRPC6 can alter podocin expression remains unknown. Western blot analysis and confocal microscopy were used to examine both expression levels of TRPC6, podocin, and nephrin and morphological changes of podocytes in response to high glucose. High glucose increased the expression of TRPC6 but reduced the expression of podocin and nephrin, in both cultured human podocytes and type 1 diabetic rat kidneys. The decreased podocin was diminished in TRPC6 knockdown podocytes. High glucose elevated intracellular Ca2+ in control podocytes but not in TRPC6 knockdown podocytes. High glucose also elevated the expression of a tight junction protein, zonula occludens-1, and induced the redistribution of zonula occludens-1 and loss of podocyte processes. These data together suggest that high glucose reduces protein levels of podocin by activating TRPC6 and induces morphological changes of cultured podocytes.
Sigma-Aldrich
Anti-TRPC6 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Nephrin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution