Merck
  • Di-2-ethylhexyl phthalate (DEHP) induces apoptosis and autophagy of mouse GC-1 spg cells.
Di-2-ethylhexyl phthalate (DEHP) induces apoptosis and autophagy of mouse GC-1 spg cells.
Environmental toxicology (2019-11-02)
Yu Gan, Dan Yang, Si Yang, Jinglei Wang, Jie Wei, Jiaxiang Chen
ABSTRACT
As a widely used plasticizer in industry, di-2-ethylhexylphthalate (DEHP) can cause testicular toxicity, yet little is known about the potential mechanism. In this study, DEHP exposure dramatically inhibited cellviability and induced apoptosis of mouse GC-1 spg cells. Furthermore, DEHP significantly increased the levels of autophagy proteins LC3-II, Beclin1 and Atg5, as well as the ratio ofLC3-II/LC3-I. Transmission electron microscopy (TEM) further confirmed that DEHP induced autophagy of mouse GC-1 spg cells. DEHP was also shown to induceoxidative stress; while inhibition of oxidative stress with NAC could increase cell viability and inhibit DEHP-induced apoptosis and autophagy. These results suggested that DEHP induced apoptosis and autophagy of mouse GC-1 spg cells via oxidative stress. 3-MA, an inhibitor of autophagy, could rescue DEHP-induced apoptosis. In summary, DEHP induced apoptosis and autophagy of mouse GC-1 spg cells via oxidative stress, and autophagy might exert a cytotoxic effect on DEHP-induced apoptosis.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard