Merck
Hybrid Systems Based on Talc and Chitosan for Controlled Drug Release.
Materials (Basel, Switzerland) (2019-11-07)
Luciano C B Lima, Caio C Coelho, Fabrícia C Silva, Andréia B Meneguin, Hernane S Barud, Roosevelt D S Bezerra, Cesar Viseras, Josy A Osajima, Edson C Silva-Filho
ABSTRACT
Inorganic matrices and biopolymers have been widely used in pharmaceutical fields. They show properties such as biocompatibility, incorporation capacity, and controlled drug release, which can become more attractive if they are combined to form hybrid materials. This work proposes the synthesis of new drug delivery systems (DDS) based on magnesium phyllosilicate (Talc) obtained by the sol-gel route method, the biopolymer chitosan (Ch), and the inorganic-organic hybrid formed between this matrix (Talc + Ch), obtained using glutaraldehyde as a crosslink agent, and to study their incorporation/release capacity of amiloride as a model drug. The systems were characterized by X-ray diffraction (XRD), Therma analysis TG/DTG, and Fourier-transform infrared spectroscopy (FTIR) that supported the DDS's formation. The hybrid showed a better drug incorporation capacity compared to the precursors, with a loading of 55.74, 49.53, and 4.71 mg g-1 for Talc + Ch, Talc, and Ch, respectively. The release assays were performed on a Hanson Research SR-8 Plus dissolver using apparatus I (basket), set to guarantee the sink conditions. The in vitro release tests showed a prolongation of the release rates of this drug for at least 4 h. This result proposes that the systems implies the slow and gradual release of the active substance, favoring the maintenance of the plasma concentration within a therapeutic window.
Sigma-Aldrich
Magnesium chloride hexahydrate, AR, ≥99%