Merck
  • Brownian-particle motion used to characterize mechanical properties of lipid vesicles.
Brownian-particle motion used to characterize mechanical properties of lipid vesicles.
The Journal of chemical physics (2020-01-10)
Idalia M Sandoval-Jiménez, Damián Jacinto-Méndez, Liliana G Toscano-Flores, Mauricio D Carbajal-Tinoco
ABSTRACT
We present a method that takes advantage of the Brownian motion of a colloidal particle to experimentally determine the area stretch modulus of giant liposomes in solution, in regard to the continuum mechanics theory of elasticity. The characteristic parameters of the corresponding model are measured by using the three-dimensional version of digital video microscopy. Such an approach makes use of the diffraction pattern generated by fluorescent spheres found below the focal plane of the microscope objective, allowing the spatial location of the sphere and, thus, the reconstruction of its trajectory. When this particle probe is localized in the neighborhood of a bilayer membrane, its motion causes an elastic distortion of the membrane that is quantifiable. More importantly, such deformation is related to the mentioned modulus, whose measured values are consistent with experimental data obtained through well-known techniques.
Product Description
Avanti
18:1-12:0 NBD PC, Avanti Research - A Croda Brand 810133P, powder
Avanti
18:1-12:0 NBD PC, Avanti Research - A Croda Brand 810133C
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
12:0 PC (DLPC), 1,2-dilauroyl-sn-glycero-3-phosphocholine, chloroform
Avanti
12:0 PC (DLPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand