Merck
  • Irradiation by γ-rays reduces the level of H3S10 phosphorylation and weakens the G2 phase-dependent interaction between H3S10 phosphorylation and γH2AX.
Irradiation by γ-rays reduces the level of H3S10 phosphorylation and weakens the G2 phase-dependent interaction between H3S10 phosphorylation and γH2AX.
Biochimie (2018-08-11)
Eva Bártová, Gabriela Lochmanová, Soňa Legartová, Jana Suchánková, Radek Fedr, Jana Krejčí, Zbyněk Zdráhal
ABSTRACT
Histone posttranslational modifications regulate diverse nuclear functions, including DNA repair. Here, we use mass spectrometry, western blotting, immunohistochemistry and advanced confocal microscopy in order to show radiation-specific changes in the histone signature. We studied wild-type mouse embryonic stem cells (mESCs) and mESCs with a depletion of histone deacetylase 1 (HDAC1), which plays a role in DNA repair. Irradiation by γ-rays increased the S139 phosphorylation of histone H2AX but reduced the level of the H3K9-R17 peptide, which contains S10 phosphorylation (H3S10ph). On an individual cellular level, H3S10ph was low in highly γH2AX-positive UV laser-induced DNA lesions, and this nuclear distribution pattern was not changed by HDAC1 depletion. Despite this fact, spontaneous γH2AX-positive DNA lesions colocalized with large H3S10ph-positive nuclear bodies that appear in the G2 phase of the cell cycle. Similarly, by FLIM-FRET analysis, we observed an interaction between H3S10ph and γH2AX in the G2 phase. However, this interaction was reduced when cells were exposed to γ-rays. A mutual link between H3S10ph and γH2AX was not observed in the G1 phase of the cell cycle. Together, our data show that despite the fact that H3S10ph is not directly involved in DNA repair, a decrease in H3S10 phosphorylation and weakened interaction between H3S10ph and γH2AX is a result of radiation-induced damage of the genome. In this case, γ-irradiation also decreased the number of cells in the G1 phase, characterized by no interaction between H3S10ph and γH2AX.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %
Sigma-Aldrich
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)