Merck
  • Genome-wide Control of Heterochromatin Replication by the Telomere Capping Protein TRF2.
Genome-wide Control of Heterochromatin Replication by the Telomere Capping Protein TRF2.
Molecular cell (2018-05-05)
Aaron Mendez-Bermudez, Liudmyla Lototska, Serge Bauwens, Marie-Josèphe Giraud-Panis, Olivier Croce, Karine Jamet, Agurtzane Irizar, Macarena Mowinckel, Stephane Koundrioukoff, Nicolas Nottet, Genevieve Almouzni, Mare-Paule Teulade-Fichou, Michael Schertzer, Mylène Perderiset, Arturo Londoño-Vallejo, Michelle Debatisse, Eric Gilson, Jing Ye
ABSTRACT
Hard-to-replicate regions of chromosomes (e.g., pericentromeres, centromeres, and telomeres) impede replication fork progression, eventually leading, in the event of replication stress, to chromosome fragility, aging, and cancer. Our knowledge of the mechanisms controlling the stability of these regions is essentially limited to telomeres, where fragility is counteracted by the shelterin proteins. Here we show that the shelterin subunit TRF2 ensures progression of the replication fork through pericentromeric heterochromatin, but not centromeric chromatin. In a process involving its N-terminal basic domain, TRF2 binds to pericentromeric Satellite III sequences during S phase, allowing the recruitment of the G-quadruplex-resolving helicase RTEL1 to facilitate fork progression. We also show that TRF2 is required for the stability of other heterochromatic regions localized throughout the genome, paving the way for future research on heterochromatic replication and its relationship with aging and cancer.
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
ICRF-193, apoptosis inducer, arabinosidase substrate
Sigma-Aldrich
VE-821, ≥98% (HPLC)
Sigma-Aldrich
KU-55933, ≥98% (HPLC)